2-Amino-1-methoxybutane ≥90.0%
Supplier: TCI America
Synonyms:
1-Methoxy-sec-butylamine
M0746-500ML
M0746-25ML
TCM0746-500MLEA
207.1
USD
TCM0746-500ML
TCM0746-025ML
2-Amino-1-methoxybutane ≥90.0%
2-Amino-1-methoxybutane
CAS Number: 63448-63-5
MDL Number: MFCD00047920
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Other Product Names / Synonyms: 1-Methoxy-sec-butylamine
Formula:
C₅H₁₃NO MW: 103.17 g/mol |
MDL Number:
MFCD00047920 CAS Number: 63448-63-5 |
Specification Test Results
Purity/Analysis Method | >90.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...