Methylcyclohexene-1,2-dicarboxylic anhydride (mixture of isomers) ≥80.0%
Supplier: TCI America
Synonyms:
Methyltetrahydrophthalic Anhydride
M0683-500G
M0683-25G
TCM0683-500GEA
109
USD
TCM0683-500G
TCM0683-025G
Methylcyclohexene-1,2-dicarboxylic anhydride (mixture of isomers) ≥80.0%
Methylcyclohexene-1,2-dicarboxylic anhydride (mixture of isomers)
CAS Number: 11070-44-3
MDL Number: MFCD00014585
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 157
Specific Gravity (20/20): 1.21
Other Product Names / Synonyms: Methyltetrahydrophthalic Anhydride
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 120 °C Density: 1.21 (20 °C) Flash Pt: 157 °C |
MDL Number:
MFCD00014585 CAS Number: 11070-44-3 |
Specification Test Results
Purity/Analysis Method | >80.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...