DL-Methionine methylsulfonium chloride ≥99.0%
Supplier: TCI America
Synonyms:
Vitamin U, (3-Amino-3-carboxypropyl)dimethylsulphonium chloride
M0644-25G
M0644-500G
TCM0644-025GEA
55
USD
TCM0644-025G
TCM0644-500G
DL-Methionine methylsulfonium chloride ≥99.0%
DL-Methionine methylsulfonium chloride
CAS Number: 3493-12-7
MDL Number: MFCD00031700
Molecular Formula: C6H14ClNO2S
Molecular Weight: 199.69
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Product Categories: Bioscience, Vitamin Related Compounds, Biologically-modified Amino Acids
Other Product Names / Synonyms: (3-Amino-3-carboxypropyl)dimethylsulfonium Chloride, S-Methylmethioninesulfonium Chloride, Vitamin U Chloride
Formula:
C₆H₁₄ClNO₂S |
MDL Number:
MFCD00031700 CAS Number: 3493-12-7 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...