4,4'-Methylenebis(2-chloroaniline) ≥90.0% (by HPLC)

Supplier: TCI America

M0609-25G M0609-500G
TCM0609-025GEA 33 USD
TCM0609-025G TCM0609-500G
4,4'-Methylenebis(2-chloroaniline) ≥90.0% (by HPLC)
4,4'-Methylenebis(2-chloroaniline)

CAS Number: 101-14-4
MDL Number: MFCD00047829
Molecular Formula: C13H12Cl2N2
Molecular Weight: 267.15
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Melting point (°C): 108
Flash Point (°C): 110


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Bis(2-chloroanilino)methane, 4,4'-Diamino-3,3'-dichlorodiphenylmethane, 3,3'-Dichloro-4,4'-diaminodiphenylmethane

Formula: C₁₃H₁₂Cl₂N₂
Melting Pt: 108 °C
Flash Pt: 110 °C
MDL Number: MFCD00047829
CAS Number: 101-14-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9a746a06e865597da7e0403193bcfb25256f901&vid=b9a746a06e865597da7e0403193bcfb25256f901&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR