2-(Morpholinothio)benzothiazole ≥97.0%

Supplier: TCI America

Synonyms: 4-(Benzo[d]thiazol-2-ylthio)morpholine, N-Oxydiethylene-2-benzothiazolesulphenamide

M0532-500G M0532-25G
TCM0532-500GEA 139 USD
TCM0532-500G TCM0532-025G
2-(Morpholinothio)benzothiazole ≥97.0%
2-(Morpholinothio)benzothiazole

CAS Number: 102-77-2
MDL Number: MFCD00022870
Molecular Formula: C11H12N2OS2
Molecular Weight: 252.35
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 86
Flash Point (°C): 195


Other Product Names / Synonyms: N-Oxydiethylene-2-benzothiazolesulfenamide

Formula: C₁₁H₁₂N₂OS₂
MW: 252.35 g/mol
Melting Pt: 86 °C
Flash Pt: 195 °C
MDL Number: MFCD00022870
CAS Number: 102-77-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2581ed25878139adfcdbe452d5d3c7066a8fa58b&vid=2581ed25878139adfcdbe452d5d3c7066a8fa58b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR