Methyl propyl ether ≥95.0%

Supplier: TCI America

M0510-25ML M0510-500ML
TCM0510-025MLEA 98 USD
TCM0510-025ML TCM0510-500ML
Methyl propyl ether ≥95.0%
Methyl propyl ether

CAS Number: 557-17-5
MDL Number: MFCD00059399
Molecular Formula: C4H10O
Molecular Weight: 74.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 39
Flash Point (°C): -20
Specific Gravity (20/20): 0.73


Other Product Names / Synonyms: 1-Methoxypropane

Formula: C₄H₁₀O
Boiling Pt: 39 °C
Density: 0.73 (20 °C)
Flash Pt: –20 °C
MDL Number: MFCD00059399
CAS Number: 557-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58f1b39f86eb47f9bbab64090ba8626cde5938be&vid=58f1b39f86eb47f9bbab64090ba8626cde5938be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR