Butyl methyl ether ≥99.0%
Supplier: TCI America
Synonyms:
1-Methoxybutane
M0504-25ML
M0504-500ML
M0504-100ML
TCM0504-025MLEA
98.67
USD
TCM0504-025ML
TCM0504-500ML
TCM0504-100ML
Butyl methyl ether ≥99.0%
Butyl methyl ether
CAS Number: 628-28-4
MDL Number: MFCD00009453
Molecular Formula: C5H12O
Molecular Weight: 88.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Flash Point (°C): -10
Specific Gravity (20/20): 0.75
Other Product Names / Synonyms: 1-Methoxybutane
Formula:
C₅H₁₂O MW: 88.15 g/mol Boiling Pt: 70 °C Density: 0.75 (20 °C) Flash Pt: –10 °C |
MDL Number:
MFCD00009453 CAS Number: 628-28-4 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...