2-Aminobenzotriazole ≥98.0%

Supplier: TCI America

Synonyms: 2-Benzotriazolamine

A1585-100MG A1585-1G
TCA1585-100MGEA 57.5 USD
TCA1585-100MG TCA1585-1G
2-Aminobenzotriazole ≥98.0%
2-Aminobenzotriazole

CAS Number: 1614-11-5
MDL Number: MFCD00724002
Molecular Formula: C6H6N4
Molecular Weight: 134.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 122


Other Product Names / Synonyms: 2-Benzotriazolamine

Formula: C₆H₆N₄
MW: 134.14 g/mol
Melting Pt: 122 °C
MDL Number: MFCD00724002
CAS Number: 1614-11-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d25d3882bf030dc87ba15a6f5a54964cc82d2589&vid=d25d3882bf030dc87ba15a6f5a54964cc82d2589&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR