Variamine blue B ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Amino-4'-methoxydiphenylamine Hydrochloride, N-(4-Methoxyphenyl)-1,4-phenylenediamine Hydrochloride, Variamine Blue B Hydrochloride

M0494-1G M0494-25G
TCM0494-001GEA 35 USD
TCM0494-001G TCM0494-025G
Variamine blue B ≥98.0% (by titrimetric analysis)
Variamine blue B

CAS Number: 3566-44-7
MDL Number: MFCD00012989
Molecular Formula: C13H14N2O
Molecular Weight: 250.73
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 255


Other Product Names / Synonyms: 4-Amino-4'-methoxydiphenylamine Hydrochloride, N-(4-Methoxyphenyl)-1,4-phenylenediamine Hydrochloride, Variamine Blue B Hydrochloride

Formula: C₁₃H₁₄N₂O
MW: 250.73 g/mol
Melting Pt: 255 °C
MDL Number: MFCD00012989
CAS Number: 3566-44-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd4a22b0f7a4cffbfa668142c87ee26e5fd8658a&vid=fd4a22b0f7a4cffbfa668142c87ee26e5fd8658a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR