5-Methyl-DL-tryptophan ≥98.0%

Supplier: TCI America

Synonyms: 2-Amino-3-(5-methyl-1H-indol-3-yl)propanoic acid, H-DL-Trp(5-Me)-OH

M0452-1G M0452-100MG
TCM0452-1GEA 564 USD
TCM0452-1G TCM0452-100MG
5-Methyl-DL-tryptophan ≥98.0%
5-Methyl-DL-tryptophan

CAS Number: 951-55-3
MDL Number: MFCD00005652
Molecular Formula: C12H14N2O2
Molecular Weight: 218.26
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal
Melting point (°C): 275
Specific rotation [a]20/D: 0 deg (C=1, HCl)


Product Categories: Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: H-DL-Trp(5-Me)-OH

Formula: C₁₂H₁₄N₂O₂
Melting Pt: 275 °C
MDL Number: MFCD00005652
CAS Number: 951-55-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a09968ae8e50f3c831287e9bb01a41eb3cb2592&vid=9a09968ae8e50f3c831287e9bb01a41eb3cb2592&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T,HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR