Sodium-N-methyltaurinate 62 - 66% in water
Supplier: TCI America
Synonyms:
Ethanesulphonic acid 2-(methylamino) sodium salt (1:1), Sodium 2-(methylamino)ethanesulphonate, N-Methyltaurine sodium salt
M0434-25G
M0434-500G
TCM0434-025GEA
51
USD
TCM0434-025G
TCM0434-500G
Sodium-N-methyltaurinate 62 - 66% in water
Sodium-N-methyltaurinate
CAS Number: 4316-74-9
MDL Number: MFCD00066598
Molecular Formula: C3H9NO3S
Molecular Weight: 161.15
Form: Clear Liquid
Color: Very Pale Yellow
Formula:
C₃H₈NNaO₃S MW: 161.1553 g/mol |
MDL Number:
MFCD00066598 CAS Number: 4316-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...