N-Methylsaccharin ≥98.0% (by GC)

Supplier: TCI America

M0427-25G M0427-5G
TCM0427-025GEA 502 USD
TCM0427-025G TCM0427-5G
N-Methylsaccharin ≥98.0% (by GC)
N-Methylsaccharin

CAS Number: 15448-99-4
MDL Number: MFCD00059263
Molecular Formula: C8H7NO3S
Molecular Weight: 197.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 131

Formula: C₈H₇NO₃S
Melting Pt: 131 °C
MDL Number: MFCD00059263
CAS Number: 15448-99-4
EINECS: 239-466-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f476dfb18b8daf21e79354ff6a72d894f33a1baf&vid=b3fa30cdd8321009ef05e5193280a934597f18a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 130.0 to 134.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR