5-Methyl-2-pyrrolidone ≥98.0%

Supplier: TCI America

M0406-10G M0406-25G
TCM0406-010GEA 52 USD
TCM0406-010G TCM0406-025G
5-Methyl-2-pyrrolidone ≥98.0%
5-Methyl-2-pyrrolidone

CAS Number: 108-27-0
MDL Number: MFCD00005273
Molecular Formula: C5H9NO
Molecular Weight: 99.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 142
Flash Point (°C): 113
Freezing point (°C): 38


Other Product Names / Synonyms: 5-Methyl-2-pyrrolidinone

Formula: C₅H₉NO
Boiling Pt: 142 °C
Flash Pt: 113 °C
MDL Number: MFCD00005273
CAS Number: 108-27-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=753658228d8c4377193f46359bb40c76f7ec57b4&vid=753658228d8c4377193f46359bb40c76f7ec57b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR