Isobutyl mercaptan ≥98.0%

Supplier: TCI America

Synonyms: 2-Methyl-1-propanethiol

M0404-25ML M0404-500ML
TCM0404-025MLEA 30.29 USD
TCM0404-025ML TCM0404-500ML
Isobutyl mercaptan ≥98.0%
Isobutyl mercaptan

CAS Number: 513-44-0
MDL Number: MFCD00004882
Molecular Formula: C4H10S
Molecular Weight: 90.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): -10
Specific Gravity (20/20): 0.83


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: 2-Methyl-1-propanethiol

Formula: C₄H₁₀S
MW: 90.18 g/mol
Boiling Pt: 88 °C
Density: 0.83 (20 °C)
Flash Pt: -10 °C
MDL Number: MFCD00004882
CAS Number: 513-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace94aef487f7aba6047a7a46349c566029915b8&vid=ace94aef487f7aba6047a7a46349c566029915b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR