DL-Menthol ≥98.0%

Supplier: TCI America

Synonyms: rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol

M0321-25ML M0321-500ML M0321-100ML
TCM0321-025MLEA 20.47 USD
TCM0321-025ML TCM0321-500ML TCM0321-100ML
DL-Menthol ≥98.0%
DL-Menthol

CAS Number: 89-78-1
MDL Number: MFCD00001484
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 85
Freezing point (°C): 28


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: (+/-)-p-Menthan-3-ol

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00001484
CAS Number: 89-78-1
EINECS: 201-939-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91fd81a8d5b9f34395e220d5a3b1244ff9f102f9&vid=91fd81a8d5b9f34395e220d5a3b1244ff9f102f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR