γ-Terpinene ≥95.0%

Supplier: TCI America
Danger

Synonyms: gamma-Terpinene, p-Mentha-1,4-diene

M0318-500ML M0318-25ML
TCM0318-500MLEA 163 USD
TCM0318-500ML TCM0318-025ML
γ-Terpinene ≥95.0%
γ-Terpinene

CAS Number: 99-85-4
MDL Number: MFCD00001537
Molecular Formula: C10H16
Molecular Weight: 136.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Flash Point (°C): 56
Specific Gravity (20/20): 0.85


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: p-Mentha-1,4-diene

Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 183 °C
Density: 0.85 (20 °C)
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001537
CAS Number: 99-85-4
EINECS: 202-794-6
UN: 2319
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dea89c2e64f2214b9d6cf778b2ff9e9328b144d8&vid=dea89c2e64f2214b9d6cf778b2ff9e9328b144d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR