α-Terpinene ≥90.0%

Supplier: TCI America

Synonyms: p-Mentha-1,3-diene, 1-Isopropyl-4-methylcyclohexa-1,3-diene

M0317-25ML M0317-500ML
TCM0317-025MLEA 41.92 USD
TCM0317-025ML TCM0317-500ML
α-Terpinene ≥90.0%
α-Terpinene

CAS Number: 99-86-5
MDL Number: MFCD00001534
Molecular Formula: C10H16
Molecular Weight: 136.24
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Flash Point (°C): 52
Specific Gravity (20/20): 0.84
Storage Temperature: 0-10°C


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: p-Mentha-1,3-diene

Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 173 °C
Melting Pt: –48 °C
Density: 0.84 (20 °C)
Flash Pt: 52 °C
Storage Temperature: Freezer
MDL Number: MFCD00001534
CAS Number: 99-86-5
EINECS: 202-795-1
UN: 2319
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f494ea3f3a3a8a64b3adc6da2cb4d2cbad20e1c8&vid=f494ea3f3a3a8a64b3adc6da2cb4d2cbad20e1c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR