Isopropyl methacrylate ≥98.0% stabilised

Supplier: TCI America

Synonyms: Methacrylic Acid Isopropyl Ester

M0280-500ML M0280-25ML
TCM0280-500MLEA 278.73 USD
TCM0280-500ML TCM0280-025ML
Isopropyl methacrylate ≥98.0% stabilised
Isopropyl methacrylate

(stabilized with MEHQ)
CAS Number: 4655-34-9
MDL Number: MFCD00048290
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 25
Specific Gravity (20/20): 0.89


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid Isopropyl Ester

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 126 °C
Density: 0.89 (20 °C)
Flash Pt: 25 °C
MDL Number: MFCD00048290
CAS Number: 4655-34-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a94d24f9908b631382a2d370a5733fdae681b844&vid=a94d24f9908b631382a2d370a5733fdae681b844&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR