Dicoumarol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M0216-1G M0216-25G
TCM0216-001GEA 35 USD
TCM0216-001G TCM0216-025G
Dicoumarol ≥98.0% (by HPLC, titration analysis)
Dicoumarol

CAS Number: 66-76-2
MDL Number: MFCD00006857
Molecular Formula: C19H12O6
Molecular Weight: 336.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 292


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: Bishydroxycoumarin, 3,3'-Methylenebis(4-hydroxycoumarin), Dicoumarin

Formula: C₁₉H₁₂O₆
Melting Pt: 292 °C
MDL Number: MFCD00006857
CAS Number: 66-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15c8fe316fe705263c13836b2bd43148f4d6bb81&vid=15c8fe316fe705263c13836b2bd43148f4d6bb81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR