1,5-Diamino-2-methylpentane ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Methyl-1,5-diaminopentane, 2-Methylpentane-1,5-diamine, 1,5-Diamino-2-methylpentane, 2-Methyl-1,5-pentanediamine
M0205-500ML
M0205-25ML
TCM0205-500MLEA
65.92
USD
TCM0205-500ML
TCM0205-025ML
1,5-Diamino-2-methylpentane ≥98.0% (by GC, titration analysis)
1,5-Diamino-2-methylpentane
CAS Number: 15520-10-2
MDL Number: MFCD00013460
Molecular Formula: C6H16N2
Molecular Weight: 116.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 85
Specific Gravity (20/20): 0.87
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,5-Diamino-2-methylpentane, 2-Methylpentamethylenediamine, 2-Methyl-1,5-pentanediamine
Formula:
C₆H₁₆N₂ MW: 116.21 g/mol Boiling Pt: 78 °C Melting Pt: –60…–50 °C Density: 0.87 (20 °C) Flash Pt: 85 °C |
MDL Number:
MFCD00013460 CAS Number: 15520-10-2 EINECS: 239-556-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...