(±)-2-Methyl-1-pentanol ≥98.0%

Supplier: TCI America

Synonyms: 2-Methylamyl alcohol, 2-Methyl-1-pentanol, 2-Propylpropanol

M0192-25ML M0192-500ML
TCM0192-025MLEA 54.91 USD
TCM0192-025ML TCM0192-500ML
(±)-2-Methyl-1-pentanol ≥98.0%
(±)-2-Methyl-1-pentanol

CAS Number: 105-30-6
MDL Number: MFCD00004745
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 41
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: 2-Methylamyl Alcohol, 2-Propylpropanol

Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 148 °C
Melting Pt: 0 °C
Density: 0.82 (20 °C)
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00004745
CAS Number: 105-30-6
EINECS: 203-285-1
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e9e676c4a34828fe2fd46a3d9c0a280ff39d08f&vid=3e9e676c4a34828fe2fd46a3d9c0a280ff39d08f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR