(±)-2-Methylbutyric acid ≥97.0%

Supplier: TCI America

Synonyms: (±)-2-Methylbutanoic acid, 2-Methylbutyric acid, 2-Methylbutanoic acid

M0181-25ML M0181-500ML
TCM0181-025MLEA 49.79 USD
TCM0181-025ML TCM0181-500ML
(±)-2-Methylbutyric acid ≥97.0%
(±)-2-Methylbutyric acid

CAS Number: 116-53-0
MDL Number: MFCD00002669
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 174
Flash Point (°C): 73
Specific Gravity (20/20): 0.94

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 174 °C
Melting Pt: –70 °C
Density: 0.94 (20 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002669
CAS Number: 116-53-0
EINECS: 204-145-2
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39989b049c18333c8b8c3fa56ece63f47a08a300&vid=39989b049c18333c8b8c3fa56ece63f47a08a300&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR