(±)-2-Methyl-1-butanol ≥97.0%

Supplier: TCI America
Danger

M0175-500ML M0175-25ML
TCM0175-500MLEA 52 USD
TCM0175-500ML TCM0175-025ML
(±)-2-Methyl-1-butanol ≥97.0%
(±)-2-Methyl-1-butanol

CAS Number: 137-32-6
MDL Number: MFCD00004743
Molecular Formula: C5H12O
Molecular Weight: 88.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 128
Flash Point (°C): 43
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: DL-sec-Butylcarbinol

Formula: C₅H₁₂O
Boiling Pt: 128 °C
Melting Pt: –70 °C
Density: 0.82 (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00004743
CAS Number: 137-32-6
EINECS: 205-289-9
UN: 1105
ADR: 3,III
Merck Index: 12,06109
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ba9ff829c3a1053a241cd3c9557201671a7327a&vid=9ba9ff829c3a1053a241cd3c9557201671a7327a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR