N-Methylbenzenesulfonamide ≥98.0%

Supplier: TCI America

M0157-25G M0157-500G
TCM0157-025GEA 33 USD
TCM0157-025G TCM0157-500G
N-Methylbenzenesulfonamide ≥98.0%
N-Methylbenzenesulphonamide

CAS Number: 5183-78-8
MDL Number: MFCD00025023
Molecular Formula: C7H9NO2S
Molecular Weight: 171.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal


Other Product Names / Synonyms: Benzenesulfonyl-N-methylamide

Formula: C₇H₉NO₂S
MDL Number: MFCD00025023
CAS Number: 5183-78-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b9f1e29f194673d91f74ab6f049099ede5aa2b5&vid=8b9f1e29f194673d91f74ab6f049099ede5aa2b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR