Methylaminoacetonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M0140-5ML M0140-25ML
TCM0140-005MLEA 138.1 USD
TCM0140-005ML TCM0140-25ML
Methylaminoacetonitrile ≥98.0% (by GC, titration analysis)
Methylaminoacetonitrile

CAS Number: 5616-32-0 MDL Number: MFCD00058964 Molecular Formula: C3H6N2 Molecular Weight: 70.10 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 65 Specific Gravity (20/20): 0.92 Storage Temperature: 0°C

Formula: C₃H₆N₂
MW: 70.10 g/mol
Boiling Pt: 65 °C
Density: 0.92 (20 °C)
MDL Number: MFCD00058964
CAS Number: 5616-32-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e3d819ae44202ff76bc1ca6c5834209397bb813&vid=1e3d819ae44202ff76bc1ca6c5834209397bb813&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR