4-Amino-N-methylphthalimide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 5-Amino-2-methylisoindoline-1,3-dione

A1561-5G A1561-25G
TCA1561-5GEA 53 USD
TCA1561-5G TCA1561-25G
4-Amino-N-methylphthalimide ≥98.0% (by HPLC, total nitrogen)
4-Amino-N-methylphthalimide

CAS Number: 2307-00-8
MDL Number: MFCD00012081
Molecular Formula: C9H8N2O2
Molecular Weight: 176.18
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 247

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
Melting Pt: 247 °C
MDL Number: MFCD00012081
CAS Number: 2307-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=774aeea34e0317d12802a26e232a134525b7970d&vid=774aeea34e0317d12802a26e232a134525b7970d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR