2-Methyl-2-propen-1-ol ≥98.0%
Supplier: TCI America
Synonyms:
3-Hydroxy-2-methylpropene, 2-Methyl-2-propen-1-ol, 2-Methyl allyl alcohol, 2-Methylprop-2-en-1-ol
M0091-25ML
M0091-500ML
M0091-5ML
TCM0091-025MLEA
78.33
USD
TCM0091-025ML
TCM0091-500ML
TCM0091-5ML
2-Methyl-2-propen-1-ol ≥98.0%
2-Methyl-2-propen-1-ol
CAS Number: 513-42-8
MDL Number: MFCD00004737
Molecular Formula: C4H8O
Molecular Weight: 72.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): 33
Specific Gravity (20/20): 0.85
Other Product Names / Synonyms: Isopropenylcarbinol, 2-Methylallyl Alcohol, 2-Methyl-2-propen-1-ol
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 113 °C Density: 0.85 (20 °C) Flash Pt: 33 °C |
MDL Number:
MFCD00004737 CAS Number: 513-42-8 EINECS: 208-161-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...