Allyl methacrylate ≥99.0% stabilised
Supplier: TCI America
Synonyms:
Methacrylic Acid Allyl Ester
M0075-25ML
M0075-500ML
TCM0075-025MLEA
26.92
USD
TCM0075-025ML
TCM0075-500ML
Allyl methacrylate ≥99.0% stabilised
Allyl methacrylate
(stabilized with MEHQ)
CAS Number: 96-05-9
MDL Number: MFCD00008592
Molecular Formula: C7H10O2
Molecular Weight: 126.16
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 64
Flash Point (°C): 35
Specific Gravity (20/20): 0.93
Product Categories: Materials Chemistry, Monomers, Monomers
Other Product Names / Synonyms: Methacrylic Acid Allyl Ester
Formula:
C₇H₁₀O₂ MW: 126.16 g/mol Boiling Pt: 64 °C Density: 0.93 (20 °C) Flash Pt: 35 °C |
MDL Number:
MFCD00008592 CAS Number: 96-05-9 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...