Methacholine chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Mecholyl Chloride, O-Acetyl-beta-methylcholine Chloride, Acetyl-β-methylcholine chloride

M0073-500G M0073-25G
TCM0073-500GEA 852 USD
TCM0073-500G TCM0073-025G
Methacholine chloride ≥98.0% (by titrimetric analysis)
Methacholine chloride

CAS Number: 62-51-1
MDL Number: MFCD00011817
Molecular Formula: C8H18ClNO2
Molecular Weight: 195.69
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 172


Other Product Names / Synonyms: O-Acetyl-beta-methylcholine Chloride, Mecholyl Chloride

Formula: C₈H₁₈ClNO₂
MW: 195.69 g/mol
Melting Pt: 172 °C
MDL Number: MFCD00011817
CAS Number: 62-51-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=312c32d701209d6337fd5ea521b177831b4bcc6e&vid=312c32d701209d6337fd5ea521b177831b4bcc6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR