3'-Amino-4'-chloroacetanilide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A1559-25G
A1559-5G
TCA1559-25GEA
125
USD
TCA1559-25G
TCA1559-5G
3'-Amino-4'-chloroacetanilide ≥98.0% (by titrimetric analysis)
3'-Amino-4'-chloroacetanilide
CAS Number: 51867-83-5
MDL Number: MFCD02093411
Molecular Formula: C8H9ClN2O
Molecular Weight: 184.62
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 172
Formula:
C₈H₉ClN₂O Melting Pt: 172 °C |
MDL Number:
MFCD02093411 CAS Number: 51867-83-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...