Loratadine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Ethyl 4-(8-chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate

L0223-100MG L0223-1G
TCL0223-100MGEA 53.47 USD
TCL0223-100MG TCL0223-1G
Loratadine ≥98.0% (by HPLC, titration analysis)
Loratadine

CAS Number: 79794-75-5
MDL Number: MFCD00672869
Molecular Formula: C22H23ClN2O2
Molecular Weight: 382.89
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 137

Formula: C₂₂H₂₃ClN₂O₂
MW: 382.88 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00672869
CAS Number: 79794-75-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a48fab5cc4caed8959d027359037aff732d6e3e9&vid=a48fab5cc4caed8959d027359037aff732d6e3e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR