D-Lysine methyl ester dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: H-D-Lys-OMe.2HCl, (R)-Methyl 2,6-diaminohexanoate dihydrochloride

L0201-5G L0201-25G
TCL0201-5GEA 127.6 USD
TCL0201-5G TCL0201-25G
D-Lysine methyl ester dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
D-Lysine methyl ester dihydrochloride

CAS Number: 67396-08-1
MDL Number: MFCD00039067
Molecular Formula: C7H16N2O2
Molecular Weight: 233.13
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 205
Specific rotation [a]20/D: -17 deg (C=5, H2O)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Lys-OMe.2HCl

Formula: C₇H₁₆N₂O₂
MW: 233.13 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00039067
CAS Number: 67396-08-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe57501e2c5966c2420c933d40e281d910e41e0d&vid=527d88a3c07c7fdc655d2707c2238f808108033c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR