Levofloxacin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (S)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid

L0193-25G L0193-5G
TCL0193-25GEA 204.7 USD
TCL0193-25G TCL0193-5G
Levofloxacin ≥98.0% (by HPLC)
Levofloxacin

CAS Number: 100986-85-4 MDL Number: MFCD00865049 Molecular Formula: C18H20FN3O4 Molecular Weight: 361.37 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Melting point (°C): 224 Specific rotation [a]20/D: -66 deg (C=5, CHCl3)

Formula: C₁₈H₂₀FN₃O₄
MW: 361.37 g/mol
Melting Pt: 224 °C
MDL Number: MFCD00865049
CAS Number: 100986-85-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6766f1c4efc0dc63a38f760f9aa24124a15327c6&vid=6766f1c4efc0dc63a38f760f9aa24124a15327c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR