Lincomycin hydrochloride monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (2S,4R)-N-((1R,2R)-2-Hydroxy-1-((2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylthio)tetrahydro-2H-pyran-2-yl)propyl)-1-methyl-4-propylpyrrolidine-2-carboxamide hydrochloride hydrate, Lincomycin HCl Monohydrate, Lincocin hydrochloride

L0166-25G L0166-5G
TCL0166-25GEA 561 USD
TCL0166-25G TCL0166-5G
Lincomycin hydrochloride monohydrate ≥98.0% (by titrimetric analysis)
Lincomycin hydrochloride monohydrate

CAS Number: 7179-49-9
MDL Number: MFCD00167245
Molecular Formula: C18H34N2O6S
Molecular Weight: 443.00
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 154
Specific rotation [a]20/D: 137 deg (C=1, H2O)


Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: Lincocin Hydrochloride

Formula: C₁₈H₃₄N₂O₆S·HCl·H₂O
MW: 461.01 g/mol
Melting Pt: 154 °C
MDL Number: MFCD00167245
CAS Number: 7179-49-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2e281544e2ee54ee3373a319085d2dbbd50554d&vid=d2e281544e2ee54ee3373a319085d2dbbd50554d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR