Lithium tetrakis(pentafluorophenyl)borate - ethyl ether complex ≥70.0%

Supplier: TCI America

L0158-1G L0158-5G
TCL0158-001GEA 291.8 USD
TCL0158-001G TCL0158-5G
Lithium tetrakis(pentafluorophenyl)borate - ethyl ether complex ≥70.0%
Lithium tetrakis(pentafluorophenyl)borate - ethyl ether complex

CAS Number: 155543-02-5 MDL Number: MFCD01861289 Molecular Formula: C24BF20Li Purity/Analysis Method: 70.0% (T) Form: Crystal

Formula: C₂₄BF₂₀Li
MDL Number: MFCD01861289
CAS Number: 155543-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c005789adc9290f6f4d491b86215e4e2c49ad66&vid=5c005789adc9290f6f4d491b86215e4e2c49ad66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 70.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR