Isopropyl linoleate 60.0%

Supplier: TCI America

Synonyms: Linoleic Acid Isopropyl Ester

L0139-25ML L0139-500ML
TCL0139-025MLEA 17 USD
TCL0139-025ML TCL0139-500ML
Isopropyl linoleate 60.0%
Isopropyl linoleate

CAS Number: 22882-95-7
MDL Number: MFCD00053809
Molecular Formula: C21H38O2
Molecular Weight: 322.53
Purity/Analysis Method: >60.0% (GC)
Form: Clear Liquid
Boiling point (°C): 179
Flash Point (°C): 191
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Linoleic Acid Isopropyl Ester

Formula: C₂₁H₃₈O₂
MW: 322.53 g/mol
Boiling Pt: 179 °C
Density: 0.87 (20 °C)
Flash Pt: 191 °C
MDL Number: MFCD00053809
CAS Number: 22882-95-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb9360f5b235320655d21063890ad2a024011684&vid=eb9360f5b235320655d21063890ad2a024011684&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >60.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR