Methyl (R)-(+)-lactate ≥98.0%

Supplier: TCI America
Danger

Synonyms: Methyl D-(+)-Lactate, (R)-Methyl 2-hydroxypropanoate, D-(+)-Lactic acid methyll ester, Methyl (R)-(+)-2-hydroxypropionate

L0136-5G L0136-25G
TCL0136-005GEA 59.57 USD
TCL0136-005G TCL0136-025G
Methyl (R)-(+)-lactate ≥98.0%
Methyl (R)-(+)-lactate

CAS Number: 17392-83-5
MDL Number: MFCD00004517
Molecular Formula: C4H8O3
Molecular Weight: 104.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 33
Flash Point (°C): 56
Specific Gravity (20/20): 1.10
Specific rotation [a]20/D: 8 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: D-(+)-Lactic Acid Methyl Ester

Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 33 °C
Density: 1.10 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00004517
CAS Number: 17392-83-5
EINECS: 241-420-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=909ad2f85765c54c4f917d9f9d55f02dddd50e7c&vid=909ad2f85765c54c4f917d9f9d55f02dddd50e7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR