4-Nitrophenyl laurate ≥98.0%
Supplier: TCI America
Synonyms:
Lauric Acid 4-Nitrophenyl Ester, p-Nitrophenyl Laurate, 4-Nitrophenyl dodecanoate
L0100-1G
L0100-5G
TCL0100-001GEA
20
USD
TCL0100-001G
TCL0100-5G
4-Nitrophenyl laurate ≥98.0%
4-Nitrophenyl laurate
CAS Number: 1956-11-2
MDL Number: MFCD00047730
Molecular Formula: C18H27NO4
Molecular Weight: 321.42
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 45
Other Product Names / Synonyms: Lauric Acid 4-Nitrophenyl Ester
Formula:
C₁₈H₂₇NO₄ MW: 321.42 g/mol Melting Pt: 45 °C |
MDL Number:
MFCD00047730 CAS Number: 1956-11-2 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...