beta-Alanyl-L-Histidine
Supplier: TCI America
A0222-1G
A0222-5G
TCA0222-001GEA
20.05
USD
TCA0222-001G
TCA0222-005G
beta-Alanyl-L-Histidine
Proteins and Peptides
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: L-Carnosine, L-Ignotine
- Melting point: 260 °C
- Specific rotation [a]20/D: 21° (C=2, H₂O)
- Solubility in water: Almost transparency
- EINECS: 206-169-9
- Merck index: 14,01850
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: L-Carnosine, L-Ignotine
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...