L-Leucine-2-naphthylamide hydrochloride ≥98.0% (by HPLC, titration analysis) 2-naphthylamine-free
Supplier: TCI America
Synonyms:
H-Leu-βNA·HCl, L-Leucine-β-naphthylamide hydrochloride
L0037-5G
L0037-1G
TCL0037-005GEA
523.1
USD
TCL0037-005G
TCL0037-001G
L-Leucine-2-naphthylamide hydrochloride ≥98.0% (by HPLC, titration analysis) 2-naphthylamine-free
L-Leucine-2-naphthylamide hydrochloride
MDL Number: MFCD00034972
Molecular Formula: C16H20N2O
Molecular Weight: 292.81
Purity/Analysis Method:
Formula:
C₁₆H₂₁ClN₂O MW: 292.807 g/mol |
MDL Number:
MFCD00034972 CAS Number: 893-36-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity(HPLC) | min. 98.0 area% |
Purity(Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +88.0 to +92.0 deg(C=1, H2O) |
Solubility in Water | almost transparency |
2-Naphthylamine | no detected |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...