N-Lauroylsarcosine sodium salt hydrate ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Danger

L0019-25G L0019-500G
TCL0019-025GEA 28.2 USD
TCL0019-025G TCL0019-500G
N-Lauroylsarcosine sodium salt hydrate ≥98.0% (by total nitrogen basis)
N-Lauroylsarcosine sodium salt hydrate

CAS Number: 137-16-6
MDL Number: MFCD00042728
Molecular Formula: C15H29NO3
Molecular Weight: 293.38
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 46


Product Categories: Materials Chemistry, Anionic Surfactants
Other Product Names / Synonyms: N-Dodecanoylsarcosine Sodium Salt, N-Lauroylsarcosine Sodium Salt, Sodium N-Dodecanoylsarcosinate

Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
Melting Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00042728
CAS Number: 137-16-6
EINECS: 205-281-5
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a2f7f10bb1daa089d95f0cf1135873c20b78a93&vid=70df086a31a17732b9f437031214dedcf9492013&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR