Ethyl lactate ≥98.0%
Supplier: TCI America
Synonyms:
2-Hydroxypropionic Acid Ethyl Ester, Ethyl 2-Hydroxypropionate, Lactic Acid Ethyl Ester
L0003-500ML
L0003-25ML
TCL0003-500MLEA
82.54
USD
TCL0003-500ML
TCL0003-025ML
Ethyl lactate ≥98.0%
Ethyl lactate
CAS Number: 97-64-3
MDL Number: MFCD00065359
Molecular Formula: C5H10O3
Molecular Weight: 118.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 154
Melting point (°C): -25
Flash Point (°C): 53
Specific Gravity (20/20): 1.03
Specific rotation [a]20/D: -10 deg
Other Product Names / Synonyms: Ethyl 2-Hydroxypropionate, 2-Hydroxypropionic Acid Ethyl Ester, Lactic Acid Ethyl Ester
Formula:
C₅H₁₀O₃ MW: 118.13 g/mol Boiling Pt: 154 °C Melting Pt: –25 °C Density: 1.03 (20 °C) Flash Pt: 53 °C |
MDL Number:
MFCD00065359 CAS Number: 97-64-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...