Khellin ≥95.0%
Supplier: TCI America
Synonyms:
Visammin, 4,9-Dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one, 4,9-Dimethoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one
K0039-5G
K0039-25G
TCK0039-005GEA
156
USD
TCK0039-005G
TCK0039-025G
Khellin ≥95.0%
Khellin
CAS Number: 82-02-0
MDL Number: MFCD00005007
Molecular Formula: C14H12O5
Molecular Weight: 260.25
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 152
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 4,9-Dimethoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one, Visammin
Formula:
C₁₄H₁₂O₅ MW: 260.25 g/mol Melting Pt: 152 °C |
MDL Number:
MFCD00005007 CAS Number: 82-02-0 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...