1-Kestose ≥99.0% (by HPLC)

Supplier: TCI America

K0032-1G K0032-5G
TCK0032-001GEA 329.49 USD
TCK0032-001G TCK0032-5G
1-Kestose ≥99.0% (by HPLC)
1-Kestose

CAS Number: 470-69-9
MDL Number: MFCD00142647
Molecular Formula: C18H32O16
Molecular Weight: 504.44
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 30 deg (C=1, H2O)

Formula: C₁₈H₃₂O₁₆
MDL Number: MFCD00142647
CAS Number: 470-69-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fcebfa30ac9e55c4a5915b9c66f1db72afd7c60&vid=8fcebfa30ac9e55c4a5915b9c66f1db72afd7c60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR