Isopropyl chloroformate ∼30% ∼2 mol/L in toluene

Supplier: TCI America

I0768-100G I0768-500G
TCI0768-100GEA 70.83 USD
TCI0768-100G TCI0768-500G
Isopropyl chloroformate ∼30% ∼2 mol/L in toluene
Isopropyl chloroformate

CAS Number: 108-23-6
MDL Number: MFCD00075068
Molecular Formula: C4H7ClO2
Molecular Weight: 122.55
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 6
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Chloroformic Acid Isopropyl Ester

Formula: C₄H₇ClO₂
Flash Pt: 6 °C
MDL Number: MFCD00075068
CAS Number: 108-23-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af9e8ea47256e4a6dd48b0c08c0d510bfd378005&vid=c33883f7500699370b51aac2516623d1ba01c8f8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR