Ibudilast ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-(2-Isopropylpyrazolo[1,5-a]pyridin-3-yl)-2-methylpropan-1-one, 3-Isobutyryl-2-isopropylpyrazolo[1,5-a]pyridine

I0740-20MG I0740-100MG
TCI0740-20MGEA 37 USD
TCI0740-20MG TCI0740-100MG
Ibudilast ≥98.0% (by HPLC, titration analysis)
Ibudilast

CAS Number: 50847-11-5
MDL Number: MFCD00864808
Molecular Formula: C14H18N2O
Molecular Weight: 230.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 175
Melting point (°C): 56
Storage Temperature: 0-10°C


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents
Other Product Names / Synonyms: 3-Isobutyryl-2-isopropylpyrazolo[1,5-a]pyridine

Formula: C₁₄H₁₈N₂O
MW: 230.31 g/mol
Boiling Pt: 175 °C
Melting Pt: 56 °C
MDL Number: MFCD00864808
CAS Number: 50847-11-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c0f77675225e6bf6599b4e566af9c0a9a99d027&vid=0c0f77675225e6bf6599b4e566af9c0a9a99d027&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR