4-Isoquinolineboronic acid pinacol ester ≥98.0%
Supplier: TCI America
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline
I0739-5G
I0739-1G
TCI0739-5GEA
277
USD
TCI0739-5G
TCI0739-1G
4-Isoquinolineboronic acid pinacol ester ≥98.0%
4-Isoquinolineboronic acid pinacol ester
CAS Number: 685103-98-4
MDL Number: MFCD05663891
Molecular Formula: C15H18BNO2
Molecular Weight: 255.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 101
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-(4-Isoquinolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Isoquinoline-4-boronic Acid Pinacol Ester
Formula:
C₁₅H₁₈BNO₂ MW: 255.1199 g/mol Melting Pt: 101 °C |
MDL Number:
MFCD05663891 CAS Number: 685103-98-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...