2-Iodobenzamide ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

I0734-25G I0734-5G
TCI0734-25GEA 152 USD
TCI0734-25G TCI0734-5G
2-Iodobenzamide ≥98.0% (by GC, total nitrogen)
2-Iodobenzamide

CAS Number: 3930-83-4
MDL Number: MFCD00099248
Molecular Formula: C7H6INO
Molecular Weight: 247.04
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 184

Formula: C₇H₆INO
MW: 247.04 g/mol
Melting Pt: 184 °C
MDL Number: MFCD00099248
CAS Number: 3930-83-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c60ab470b757321c5cbf970cc82998315c31744&vid=2c60ab470b757321c5cbf970cc82998315c31744&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR