Itraconazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
I0732-5G
I0732-25G
TCI0732-5GEA
138.98
USD
TCI0732-5G
TCI0732-25G
Itraconazole ≥98.0% (by HPLC, titration analysis)
Itraconazole
CAS Number: 84625-61-6
MDL Number: MFCD00870168
Molecular Formula: C35H38Cl2N8O4
Molecular Weight: 705.64
Purity/Analysis Method:
Formula:
C₃₅H₃₈Cl₂N₈O₄ Melting Pt: 169 °C |
MDL Number:
MFCD00870168 CAS Number: 84625-61-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...