4-Isopropenyltoluene ≥95.0% stabilized

Supplier: TCI America

Synonyms: 4,alpha-Dimethylstyrene, 1-Isopropenyl-4-methylbenzene

I0693-25G I0693-5G
TCI0693-25GEA 463 USD
TCI0693-25G TCI0693-5G
4-Isopropenyltoluene ≥95.0% stabilized
4-Isopropenyltoluene

(stabilized with TBC)
CAS Number: 1195-32-0
MDL Number: MFCD00036510
Molecular Formula: C10H12
Molecular Weight: 132.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 186
Melting point (°C): -28
Flash Point (°C): 62
Specific Gravity (20/20): 0.91
Storage Temperature: <0°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1-Isopropenyl-4-methylbenzene, 4,alpha-Dimethylstyrene

Formula: C₁₀H₁₂
MW: 132.21 g/mol
Boiling Pt: 186 °C
Melting Pt: –28 °C
Density: 0.91 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00036510
CAS Number: 1195-32-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85621ffdb9a0d3ac7caf99a2add8dc84df7a09f1&vid=85621ffdb9a0d3ac7caf99a2add8dc84df7a09f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR